Methyl 2-bromo-3-chlorobenzoate
Lieferant: Apollo Scientific
|
Achtung
|
OR400105-1G
OR400105-5G
OR400105-25G
APOSOR400105-1GEA
60.9
EUR
APOSOR400105-1G
APOSOR400105-5G
APOSOR400105-25G
Methyl 2-bromo-3-chlorobenzoate
Methyl 2-bromo-3-chlorobenzoate
Formel:
C₈H₆BrClO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18398719 CAS-Nummer: 1107627-14-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...