4-Bromo-5-methylbenzene-1,2-diamine
Lieferant: Apollo Scientific
OR400118-5G
APOSOR400118-5GEA
60.9
EUR
APOSOR400118-5G
4-Bromo-5-methylbenzene-1,2-diamine
4-Bromo-5-methylbenzene-1,2-diamine
Formel:
C₇H₉BrN₂ Molecular Weight: 201.07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16987757 CAS-Nummer: 102169-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...