5-Bromo-6-methyl-1H-benzimidazole 97%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-6-methyl-1H-benzo[d]imidazole

OR400129-250MG
APOSOR400129-250MGEA 112 EUR
APOSOR400129-250MG
5-Bromo-6-methyl-1H-benzimidazole 97%
5-Bromo-6-methyl-1H-benzimidazole
Formel: C₈H₇BrN₂
Molecular Weight: 211,059 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11846340
CAS-Nummer: 116106-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR