2-Iodophenacyl bromide 95%

Lieferant: Apollo Scientific

OR400132-1G OR400132-250MG OR400132-5G
APOSOR400132-1GEA 0 EUR
APOSOR400132-1G APOSOR400132-250MG APOSOR400132-5G
2-Iodophenacyl bromide 95%
2-Iodophenacyl bromide
Formel: C₈H₆BrIO
Lagertemperatur: Raumtemperatur
MDL: MFCD13194409
CAS-Nummer: 113258-84-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR