4-Methoxy-3-nitrophenol

Lieferant: Apollo Scientific
Achtung

OR400153-1G OR400153-5G
APOSOR400153-1GEA 78.3 EUR
APOSOR400153-1G APOSOR400153-5G
4-Methoxy-3-nitrophenol
4-Methoxy-3-nitrophenol
Formel: C₇H₇NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00031284
CAS-Nummer: 15174-02-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR