4-Methoxy-3-nitrophenol
Lieferant: Apollo Scientific
|
Achtung
|
OR400153-1G
OR400153-5G
APOSOR400153-1GEA
78.3
EUR
APOSOR400153-1G
APOSOR400153-5G
4-Methoxy-3-nitrophenol
4-Methoxy-3-nitrophenol
Formel:
C₇H₇NO₄ Molecular Weight: 169.135 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00031284 CAS-Nummer: 15174-02-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...