2-Amino-3-chlorphenol
Lieferant: Apollo Scientific
|
Achtung
|
OR400160-5G
APOSOR400160-5GEA
186
EUR
APOSOR400160-5G
2-Amino-3-chlorphenol
2-Amino-3-chlorphenol
Formel:
C₆H₆ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD11848418 CAS-Nummer: 56962-00-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...