4-Bromo-3-methoxybenzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Bromo-3-methoxyphenyl)methanol

OR400176-1G
APOSOR400176-1GEA 72.5 EUR
APOSOR400176-1G
4-Bromo-3-methoxybenzyl alcohol
4-Bromo-3-methoxybenzyl alcohol
Formel: C₈H₉BrO₂
Molecular Weight: 217,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11053671
CAS-Nummer: 17100-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR