Methyl 3-bromo-5-methylbenzoate

Lieferant: Apollo Scientific

OR400190-1G
APOSOR400190-1GEA 26.1 EUR
APOSOR400190-1G
Methyl 3-bromo-5-methylbenzoate
Methyl 3-bromo-5-methylbenzoate
Formel: C₉H₉BrO₂
Molecular Weight: 229.071 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11007827
CAS-Nummer: 478375-40-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR