4-Bromo-5-methyl-2-nitrophenol

Lieferant: Apollo Scientific

OR400198-1G
APOSOR400198-1GEA 136 EUR
APOSOR400198-1G
4-Bromo-5-methyl-2-nitrophenol
4-Bromo-5-methyl-2-nitrophenol
Formel: C₇H₆BrNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12923347
CAS-Nummer: 182500-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR