2-Chloro-5-iodobenzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
OR400200-1G
OR400200-5G
OR400200-25G
APOSOR400200-1GEA
0
EUR
APOSOR400200-1G
APOSOR400200-5G
APOSOR400200-25G
2-Chloro-5-iodobenzyl bromide
2-Chloro-5-iodobenzyl bromide
Formel:
C₇H₅BrClI Schmelzpunkt: 94…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08236727 CAS-Nummer: 793695-85-9 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...