2-Chloro-5-iodobenzyl bromide

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-(Bromomethyl)-1-chloro-4-iodobenzene

OR400200-1G OR400200-5G OR400200-25G
APOSOR400200-1GEA 26.1 EUR
APOSOR400200-1G APOSOR400200-5G APOSOR400200-25G
2-Chloro-5-iodobenzyl bromide
2-Chloro-5-iodobenzyl bromide
Formel: C₇H₅BrClI
Molecular Weight: 331.376 g/mol
Schmelzpunkt: 94…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08236727
CAS-Nummer: 793695-85-9
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR