Methyl-2-chlor-5-iodbenzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-chloro-5-iodobenzoate

OR400205-1G OR400205-5G OR400205-25G
APOSOR400205-1GEA 21.9 EUR
APOSOR400205-1G APOSOR400205-5G APOSOR400205-25G
Methyl-2-chlor-5-iodbenzoat
Methyl-2-chlor-5-iodbenzoat
Formel: C₈H₆ClIO₂
Molecular Weight: 296.4900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07779253
CAS-Nummer: 620621-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR