3-Bromo-5-methylbenzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
OR400207-1G
OR400207-250MG
OR400207-5G
APOSOR400207-1GEA
0
EUR
APOSOR400207-1G
APOSOR400207-250MG
APOSOR400207-5G
3-Bromo-5-methylbenzyl bromide
3-Bromo-5-methylbenzyl bromide
Formel:
C₈H₈Br₂ Schmelzpunkt: 58…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD18204792 CAS-Nummer: 51719-69-8 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...