2'-Brom-4'-chloracetophenon

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Bromo-4-chlorophenyl)ethanone

OR400212-5G
APOSOR400212-5GEA 85.7 EUR
APOSOR400212-5G
2'-Brom-4'-chloracetophenon
2'-Brom-4'-chloracetophenon
Formel: C₈H₆BrClO
Molecular Weight: 233,4896 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02683063
CAS-Nummer: 825-40-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR