3-(Methylsulphonyl)benzyl bromide 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR40022-1G
OR40022-250MG
OR40022-5G
APOSOR40022-1GEA
0
EUR
APOSOR40022-1G
APOSOR40022-250MG
APOSOR40022-5G
3-(Methylsulphonyl)benzyl bromide 97%
3-(Methylsulphonyl)benzyl bromide
Formel:
C₈H₉BrO₂S Schmelzpunkt: 72…74 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD12025160 CAS-Nummer: 82657-76-9 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...