3-Methoxy-4-nitrobenzyl bromide

Lieferant: Apollo Scientific
Gefahr

OR400222-250MG OR400222-1G
APOSOR400222-250MGEA 0 EUR
APOSOR400222-250MG APOSOR400222-1G
3-Methoxy-4-nitrobenzyl bromide
3-Methoxy-4-nitrobenzyl bromide
Formel: C₈H₈BrNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08236862
CAS-Nummer: 23145-65-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR