6-Iodo-1,3-benzoxazole 95%

Lieferant: Apollo Scientific
Achtung

OR400224-1G
APOSOR400224-1GEA 955 EUR
APOSOR400224-1G
6-Iodo-1,3-benzoxazole 95%
6-Iodo-1,3-benzoxazole
Formel: C₇H₄INO
Molecular Weight: 245,02 g/mol
MDL: MFCD28014072
CAS-Nummer: 1823954-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR