4-Methoxypyrimidin-2-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4-methoxypyrimidine

OR40024-1G OR40024-250MG
APOSOR40024-1GEA 24.8 EUR
APOSOR40024-1G APOSOR40024-250MG
4-Methoxypyrimidin-2-amine
4-Methoxypyrimidin-2-amine
Formel: C₅H₇N₃O
Lagertemperatur: Kühlschrank
MDL: MFCD00619233
CAS-Nummer: 155-90-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR