Methyl 2-ethynylbenzoate 95%

Lieferant: Apollo Scientific

OR400248-1G
APOSOR400248-1GEA 108 EUR
APOSOR400248-1G
Methyl 2-ethynylbenzoate 95%
Methyl 2-ethynylbenzoate
Formel: C₁₀H₈O₂
Molecular Weight: 160.169 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12547057
CAS-Nummer: 33577-99-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR