4-Ethinylbenzonitril

Lieferant: Apollo Scientific
Achtung

OR400253-1G OR400253-5G OR400253-25G
APOSOR400253-1GEA 79.9 EUR
APOSOR400253-1G APOSOR400253-5G APOSOR400253-25G
4-Ethinylbenzonitril
4-Ethinylbenzonitril
Formel: C₉H₅N
Lagertemperatur: Raumtemperatur
MDL: MFCD04974058
CAS-Nummer: 3032-92-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR