2-Chloro-7-methoxy-3-quinolinecarbaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-7-methoxyquinoline-3-carbaldehyde

OR40027-1G
APOSOR40027-1GEA 40.6 EUR
APOSOR40027-1G
2-Chloro-7-methoxy-3-quinolinecarbaldehyde
2-Chloro-7-methoxy-3-quinolinecarbaldehyde
Formel: C₁₁H₈ClNO₂
Molecular Weight: 221,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02227048
CAS-Nummer: 68236-20-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR