4-Methoxy-3,5-dimethylbenzaldehyde

Lieferant: Apollo Scientific

OR400287-5G
APOSOR400287-5GEA 87 EUR
APOSOR400287-5G
4-Methoxy-3,5-dimethylbenzaldehyde
4-Methoxy-3,5-dimethylbenzaldehyde
Formel: C₁₀H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02261755
CAS-Nummer: 39250-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR