4-Ethinylanilin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Aminophenyl)acetylen, 1-Ethinyl-4-aminobenzol, (4-Aminophenyl)ethin, 1-Amino-4-ethinylbenzol
OR400295-25G
APOSOR400295-25GEA
106
EUR
APOSOR400295-25G
4-Ethinylanilin 98%
4-Ethinylanilin
Formel:
C₈H₇N Molecular Weight: 117,15 g/mol Schmelzpunkt: 100…102 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00168824 CAS-Nummer: 14235-81-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...