2-Bromo-5-cyanobenzoic acid

Lieferant: Apollo Scientific
Achtung

OR400316-100MG
APOSOR400316-100MGEA 43.5 EUR
APOSOR400316-100MG
2-Bromo-5-cyanobenzoic acid
2-Bromo-5-cyanobenzoic acid
Formel: C₈H₄BrNO₂
Molecular Weight: 226.027 g/mol
Schmelzpunkt: 183…185 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11847005
CAS-Nummer: 845616-12-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR