3'-Chloro-5'-methoxyacetophenone

Lieferant: Apollo Scientific
Achtung

OR400320-1G OR400320-250MG OR400320-5G
APOSOR400320-1GEA 406 EUR
APOSOR400320-1G APOSOR400320-250MG APOSOR400320-5G
3'-Chloro-5'-methoxyacetophenone
3'-Chloro-5'-methoxyacetophenone
Formel: C₉H₉ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22544395
CAS-Nummer: 214760-34-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR