3'-Chloro-5'-methoxyacetophenone
Lieferant: Apollo Scientific
|
Achtung
|
OR400320-1G
OR400320-250MG
OR400320-5G
APOSOR400320-1GEA
406
EUR
APOSOR400320-1G
APOSOR400320-250MG
APOSOR400320-5G
3'-Chloro-5'-methoxyacetophenone
3'-Chloro-5'-methoxyacetophenone
Formel:
C₉H₉ClO₂ Molecular Weight: 184.62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22544395 CAS-Nummer: 214760-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...