3-Chloro-5-methoxybenzyl bromide

Lieferant: Apollo Scientific
Gefahr

OR400328-1G OR400328-250MG OR400328-5G
APOSOR400328-1GEA 0 EUR
APOSOR400328-1G APOSOR400328-250MG APOSOR400328-5G
3-Chloro-5-methoxybenzyl bromide
3-Chloro-5-methoxybenzyl bromide
Formel: C₈H₈BrClO
Lagertemperatur: Raumtemperatur
MDL: MFCD08234654
CAS-Nummer: 885270-32-6
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR