(4S)-2,2-Dioxido-4- isopropyl -1,2,3-oxathiazolidine, N-BOC protected
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-tert-Butyl 4-isopropyl-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide
OR400335-1G
OR400335-250MG
OR400335-5G
APOSOR400335-1GEA
212
EUR
APOSOR400335-1G
APOSOR400335-250MG
APOSOR400335-5G
(4S)-2,2-Dioxido-4- isopropyl -1,2,3-oxathiazolidine, N-BOC protected
(4S)-2,2-Dioxido-4- isopropyl -1,2,3-oxathiazolidine, N-BOC protected
Formel:
C₁₀H₁₉NO₅S Molecular Weight: 265.327 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD17018789 CAS-Nummer: 1206227-46-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...