6-Bromoisobenzofuran-1(3H)-one

Lieferant: Apollo Scientific

Synonyme: 6-Bromophthalide

OR400356-5G
APOSOR400356-5GEA 85.7 EUR
APOSOR400356-5G
6-Bromoisobenzofuran-1(3H)-one
6-Bromoisobenzofuran-1(3H)-one
Formel: C₈H₅BrO₂
Molecular Weight: 213.0281 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03428561
CAS-Nummer: 19477-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR