5-Chloro-3-iodoisatoic anhydride

Lieferant: Apollo Scientific
Achtung

OR400378-1G OR400378-250MG OR400378-5G
APOSOR400378-1GEA 0 EUR
APOSOR400378-1G APOSOR400378-250MG APOSOR400378-5G
5-Chloro-3-iodoisatoic anhydride
5-Chloro-3-iodoisatoic anhydride
Formel: C₈H₃ClINO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28154805
CAS-Nummer: 33115-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR