7-Bromo-5-chloro-1H-benzimidazole
Lieferant: Apollo Scientific
Synonyme:
7-Bromo-5-chloro-1H-benzo[d]imidazole
OR400382-1G
APOSOR400382-1GEA
183
EUR
APOSOR400382-1G
7-Bromo-5-chloro-1H-benzimidazole
7-Bromo-5-chloro-1H-benzimidazole
Formel:
C₇H₄BrClN₂ Molecular Weight: 231,477 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22558144 CAS-Nummer: 1360934-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...