3-Bromo-5-chlorobenzene -1,2-diamine 98%

Lieferant: Apollo Scientific

OR400386-1G
APOSOR400386-1GEA 230 EUR
APOSOR400386-1G
3-Bromo-5-chlorobenzene -1,2-diamine 98%
3-Bromo-5-chlorobenzene -1,2-diamine
Formel: C₆H₆BrClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD15144647
CAS-Nummer: 500862-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR