4-Bromo-3-chlorobenzene-1,2-diamine 95%

Lieferant: Apollo Scientific

OR400387-1G
APOSOR400387-1GEA 128 EUR
APOSOR400387-1G
4-Bromo-3-chlorobenzene-1,2-diamine 95%
4-Bromo-3-chlorobenzene-1,2-diamine
Formel: C₆H₆BrClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22384798
CAS-Nummer: 1008361-80-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR