3-Amino-4-bromphenol 98%

Lieferant: Apollo Scientific
Achtung

OR400395-1G
APOSOR400395-1GEA 29 EUR
APOSOR400395-1G
3-Amino-4-bromphenol 98%
3-Amino-4-bromphenol
Formel: C₆H₆BrNO
Molecular Weight: 188,0200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09260886
CAS-Nummer: 100367-37-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR