4-Amino-3-iodophenol
Lieferant: Apollo Scientific
|
Achtung
|
OR400417-1G
OR400417-250MG
APOSOR400417-1GEA
364
EUR
APOSOR400417-1G
APOSOR400417-250MG
4-Amino-3-iodophenol
4-Amino-3-iodophenol
Formel:
C₆H₆INO Molecular Weight: 235.022 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16999396 CAS-Nummer: 66416-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...