4-Iodo-3-methoxyaniline 95+%

Lieferant: Apollo Scientific

OR400428-250MG
APOSOR400428-250MGEA 0 EUR
APOSOR400428-250MG
4-Iodo-3-methoxyaniline 95+%
4-Iodo-3-methoxyaniline
Formel: C₇H₈INO
Lagertemperatur: Raumtemperatur
MDL: MFCD11041297
CAS-Nummer: 1112840-98-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR