2-Oxo-2,3-dihydro-1,3-benzoxazole-6-carboxylic acid 95+%

Lieferant: Apollo Scientific

Synonyme: 2-Oxo-2,3-dihydrobenzo[d]oxazole-6-carboxylic acid

OR400444-250MG
APOSOR400444-250MGEA 46.4 EUR
APOSOR400444-250MG
2-Oxo-2,3-dihydro-1,3-benzoxazole-6-carboxylic acid 95+%
2-Oxo-2,3-dihydro-1,3-benzoxazole-6-carboxylic acid
Formel: C₈H₅NO₄
Molecular Weight: 179,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11179328
CAS-Nummer: 54903-16-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR