6-Chloro-1,3-benzoxazole

Lieferant: Apollo Scientific

Synonyme: 6-Chlorobenzoxazole

OR400446-5G
APOSOR400446-5GEA 215 EUR
APOSOR400446-5G
6-Chloro-1,3-benzoxazole
6-Chloro-1,3-benzoxazole
Formel: C₇H₄ClNO
Molecular Weight: 153,566 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027437
CAS-Nummer: 227197-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR