Methyl 2-chloro-1,3-benzoxazole-6-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

OR400449-250MG
APOSOR400449-250MGEA 567 EUR
APOSOR400449-250MG
Methyl 2-chloro-1,3-benzoxazole-6-carboxylate 95%
Methyl 2-chloro-1,3-benzoxazole-6-carboxylate
Formel: C₉H₆ClNO₃
Molecular Weight: 211,602 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD20528144
CAS-Nummer: 819076-91-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR