4-Bromo-3-iodobenzonitrile

Lieferant: Apollo Scientific

OR400457-1G
APOSOR400457-1GEA 134 EUR
APOSOR400457-1G
4-Bromo-3-iodobenzonitrile
4-Bromo-3-iodobenzonitrile
Formel: C₇H₃BrIN
Molecular Weight: 307,914 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18391645
CAS-Nummer: 1006715-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR