Methyl 3-amino-5-bromobenzo[b]thiophene-2-carboxylate

Lieferant: Apollo Scientific

OR400462-1G
APOSOR400462-1GEA 121 EUR
APOSOR400462-1G
Methyl 3-amino-5-bromobenzo[b]thiophene-2-carboxylate
Methyl 3-amino-5-bromobenzo[b]thiophene-2-carboxylate
Formel: C₁₀H₈BrNO₂S
Molecular Weight: 286,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13689055
CAS-Nummer: 1036380-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR