3-Benzyloxybenzylbromid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-(Benzyloxy)benzyl bromide, 1-(Benzyloxy)-3-(bromomethyl)benzene

OR400465-1G
APOSOR400465-1GEA 21.9 EUR
APOSOR400465-1G
3-Benzyloxybenzylbromid
3-Benzyloxybenzylbromid
Formel: C₁₄H₁₃BrO
Molecular Weight: 277,1600 g/mol
Schmelzpunkt: 54…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318307
CAS-Nummer: 1700-31-8
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR