2-(Pyridin-4-yl)ethane-1-sulphonyl chloride
Lieferant: Apollo Scientific
OR400473-1G
APOSOR400473-1GEA
451
EUR
APOSOR400473-1G
2-(Pyridin-4-yl)ethane-1-sulphonyl chloride
2-(Pyridin-4-yl)ethane-1-sulphonyl chloride
Formel:
C₇H₈ClNO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD08445803 CAS-Nummer: 252670-82-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...