3-(Benzyloxy)benzoesäure

Lieferant: Apollo Scientific

Synonyme: 3-Benzyloxybenzoic acid

OR400474-5G
APOSOR400474-5GEA 63.8 EUR
APOSOR400474-5G
3-(Benzyloxy)benzoesäure
3-(Benzyloxy)benzoesäure
Formel: C₁₄H₁₂O₃
Molecular Weight: 228.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00527901
CAS-Nummer: 69026-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR