4-(Benzyloxy)benzyl bromide

Lieferant: Apollo Scientific

OR400483-1G OR400483-250MG
APOSOR400483-1GEA 0 EUR
APOSOR400483-1G APOSOR400483-250MG
4-(Benzyloxy)benzyl bromide
4-(Benzyloxy)benzyl bromide
Formel: C₁₄H₁₃BrO
Lagertemperatur: Raumtemperatur
MDL: MFCD07368336
CAS-Nummer: 5544-60-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR