5-Hydroxypyrazine-2-carboxylic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Oxo-4,5-dihydropyrazine-2-carboxylic acid

OR40051-100G OR40051-25G
APOSOR40051-100GEA 209 EUR
APOSOR40051-100G APOSOR40051-25G
5-Hydroxypyrazine-2-carboxylic acid 98%
5-Hydroxypyrazine-2-carboxylic acid
Formel: C5H4N2O3
Molecular Weight: 140.1000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11978170
CAS-Nummer: 34604-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR