Methyl-3-amino-6-brom-2-pyrazincarboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-amino-6-bromopyrazine-2-carboxylate
OR40052-1G
APOSOR40052-1GEA
21.9
EUR
APOSOR40052-1G
Methyl-3-amino-6-brom-2-pyrazincarboxylat
Methyl-3-amino-6-bromopyrazine-2-carboxylate
Formel:
C₆H₆BrN₃O₂ Molecular Weight: 232.03 g/mol Schmelzpunkt: 175…176 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00834964 CAS-Nummer: 6966-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...