6-Chlor-2-pyrazincarbonsäure

Lieferant: Apollo Scientific
Achtung

OR40053-1G
APOSOR40053-1GEA 21.9 EUR
APOSOR40053-1G
6-Chlor-2-pyrazincarbonsäure
6-Chloropyrazine-2-carboxylic acid
Formel: C₅H₃ClN₂O₂
Molecular Weight: 158,54 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00837871
CAS-Nummer: 23688-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR