N-[2-(Diethylamino)ethyl]-2,4-dimethyl-5-formyl-1H-pyrrole-3-carboxamide 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-{[2-(Diethylamino)ethyl]carbamoyl}-2,4-dimethyl-1H-pyrrole-2-carboxaldehyde, N-(2-(Diethylamino)ethyl)-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxamide, 3-{[2-(Diethylamino)ethyl]carbamoyl}-2,4-dimethyl-5-formyl-1H-pyrrole

OR40055-25G
APOSOR40055-25GEA 91.5 EUR
APOSOR40055-25G
N-[2-(Diethylamino)ethyl]-2,4-dimethyl-5-formyl-1H-pyrrole-3-carboxamide 98%
N-[2-(Diethylamino)ethyl]-2,4-dimethyl-5-formyl-1H-pyrrole-3-carboxamide
Formel: C₁₄H₂₃N₃O₂
Molecular Weight: 265,351 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12913910
CAS-Nummer: 356068-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR